missing translation for 'onlineSavingsMsg'
Learn More

Diphenyleneiodonium chloride, 97+%

Catalog No. AC328660250
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328660250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC328660250 Supplier Thermo Scientific Chemicals Supplier No. 328660250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4673-26-1 | C12H8ClI | 314.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4673-26-1
Molecular Formula C12H8ClI
Molecular Weight (g/mol) 314.55
MDL Number MFCD00214165
InChI Key FCFZKAVCDNTYID-UHFFFAOYSA-M
Synonym diphenyleneiodonium chloride, dibenziodolium chloride, unii-7m9d81yz2n, dpi, 8? 3-iodatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2,4,6,9,11-hexaen-8-ylium chloride, dibenziodolium, chloride, d0n0gj, c12h8i.cl, dibenzo b,d iodolium chloride, 2,2'-biphenylyleneiodonium chloride
PubChem CID 2733504
ChEBI CHEBI:77967
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3[I+]2.[Cl-]

Specifications

Melting Point 312.0°C to 322.0°C
Color White
Assay Percent Range 97+%
Quantity 25 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in dmso
Formula Weight 314.55
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material Diphenyleneiodonium chloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only