missing translation for 'onlineSavingsMsg'
Learn More

Diphenyldichlorogermane 95.0+%, TCI America™

Catalog No. D18241G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D18241G Supplier TCI America Supplier No. D18241G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1613-66-7
Molecular Formula C12H14Cl2Ge
Molecular Weight (g/mol) 301.77
MDL Number MFCD00013587
InChI Key YROQISGFJISKEN-UHFFFAOYSA-N
Synonym diphenyldichlorogermane, diphenylgermanium dichloride, germane, dichlorodiphenyl, dichlorodiphenylgermene, dichloro diphenyl germane, acmc-1bssq, germane,dichlorodiphenyl, germanium diphenyl dichloride
PubChem CID 74167
IUPAC Name dichloro(diphenyl)germane
SMILES Cl.Cl.[GeH2](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 9°C
Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 297.74
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diphenyldichlorogermane