missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine, 1% (w/v) in Sulfuric Acid, Ricca Chemical

Catalog No. 25844
Change view
Click to view available options
Quantity:
120 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
25844 120 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25844 Supplier Ricca Chemical Company Supplier No. 25844

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5 , 122-39-4
Molecular Formula C12H11N
Molecular Weight (g/mol) Mixture
InChI Key DMBHHRLKUKUOEG-UHFFFAOYSA-N
Synonym diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
PubChem CID 11487
ChEBI CHEBI:4640
IUPAC Name N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=CC=CC=C2

Specifications

CAS Min % 97.0, 0.53
CAS Max % 100.0, 0.55
Color Colorless
Packaging Amber Glass Bottle
Quantity 120 mL
Percent Purity 1% w/v in Sulfuric Acid
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1% (w/v) in Sulfuric Acid
Physical Form Liquid
Chemical Name or Material Diphenylamine
ShelfLife 12 months