missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine, 0.3% (w/v) in 50% (v/v) Sulfuric Acid, Ricca Chemical

Supplier:  Ricca Chemical Company 258616

Quantity 16 oz.
Packaging Safety-coated Glass Bottle

 View more versions of this product

Catalog No. 258616

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

122-39-4 , 7664-93-9 , 7732-18-5
169.23
diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
CHEBI:4640
N(C1=CC=CC=C1)C1=CC=CC=C1
C12H11N
DMBHHRLKUKUOEG-UHFFFAOYSA-N
11487  
N-phenylaniline

Specifications

Diphenylamine
0.15, 46.89, 51.0
16 oz.
diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
N(C1=CC=CC=C1)C1=CC=CC=C1
169.23
CHEBI:4640
Safety-coated Glass Bottle
122-39-4 , 7664-93-9 , 7732-18-5
0.16, 48.8, 53.0
C12H11N
DMBHHRLKUKUOEG-UHFFFAOYSA-N
N-phenylaniline
11487  
0.3% (w/v) in 50% (v/v) Sulfuric Acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 months

SDS
Documents

Documents