missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Phthalate 98.0+%, TCI America™

Catalog No. P030525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P030525G 25 g
P0305500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P030525G Supplier TCI America Supplier No. P030525G

Chemical Identifiers

CAS 84-62-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00003038
InChI Key DWNAQMUDCDVSLT-UHFFFAOYSA-N
Synonym diphenyl phthalate, phenyl phthalate, phthalic acid, diphenyl ester, diphenylphthalate, 1,2-benzenedicarboxylic acid, diphenyl ester, phthalic acid diphenyl ester, caswell no. 399b, unii-bu20109xhv, 1,2-benzenedicarboxylic acid, 1,2-diphenyl ester, epa pesticide chemical code 017001
PubChem CID 6778
ChEBI CHEBI:60819
IUPAC Name 1,2-diphenyl benzene-1,2-dicarboxylate
SMILES O=C(OC1=CC=CC=C1)C1=CC=CC=C1C(=O)OC1=CC=CC=C1

Specifications

Melting Point 75°C
Color White
Boiling Point 257°C
Quantity 25 g
Formula Weight 318.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl Phthalate
EINECSNumber (3)-3592
RTECSNumber TI1935000
TSCA Yes