missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl phosphate, 97%

Catalog No. AAL0212106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0212106 5 g
AAL0212114 25 g
AAL0212122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0212106 Supplier Thermo Scientific Chemicals Supplier No. L0212106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 838-85-7 | C12H11O4P | 250.19 g/mol

Diphenyl phosphate is used in the manufacture of phosphoric acid diesters such as triphenyl phosphate, trixylenyl phosphate, isodecyl diphenyl phosphate, cresyl diphenyl phosphate and isopropylphenyl diphenyl phosphate. It is widely used as a protective agent for hydroxyl group in organic synthesis. It finds application as an additive for paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl phosphate is used in the manufacture of phosphoric acid diesters such as triphenyl phosphate, trixylenyl phosphate, isodecyl diphenyl phosphate, cresyl diphenyl phosphate and isopropylphenyl diphenyl phosphate. It is widely used as a protective agent for hydroxyl group in organic synthesis. It finds application as an additive for paints and coatings.

Solubility
Soluble in benzene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 838-85-7
Molecular Formula C12H11O4P
Molecular Weight (g/mol) 250.19
MDL Number MFCD00003033
InChI Key ASMQGLCHMVWBQR-UHFFFAOYSA-N
Synonym diphenyl phosphate, phosphoric acid, diphenyl ester, phenyl hydrogen phosphate, phenyl phosphate pho 2 ho po, diphenoxyphosphinic acid, phosphoric acid diphenyl ester, diphenylphosphoric acid, phosphoric acid diphenyl, dsstox_cid_28182
PubChem CID 13282
IUPAC Name diphenyl hydrogen phosphate
SMILES OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Melting Point 62°C to 70°C
Quantity 5 g
Beilstein 1379164
Solubility Information Soluble in benzene. Insoluble in water.
Formula Weight 250.19
Percent Purity 97%
Chemical Name or Material Diphenyl phosphate
EINECSNumber 212-657-2
RTECSNumber TC5470000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only