missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Isophthalate 99.0+%, TCI America™

Catalog No. I041625G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I041625G 25 g
I041610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I041625G Supplier TCI America Supplier No. I041625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 744-45-6
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.328
MDL Number MFCD00046051
InChI Key FHESUNXRPBHDQM-UHFFFAOYSA-N
Synonym diphenyl isophthalate, diphenyl m-phthalate, diphenylisophthalate, unii-wh4w31qij8, 1,3-benzenedicarboxylic acid, diphenyl ester, isophthalic acid diphenyl ester, isophthalic acid, diphenyl ester, wh4w31qij8, 1,3-benzenedicarboxylic acid, 1,3-diphenyl ester, 1,3-diphenyl benzene-1,3-dicarboxylate
PubChem CID 69779
IUPAC Name diphenyl benzene-1,3-dicarboxylate
SMILES C1=CC=C(C=C1)OC(=O)C2=CC(=CC=C2)C(=O)OC3=CC=CC=C3

Specifications

Melting Point 138°C
Color White
Quantity 25 g
Formula Weight 318.33
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl Isophthalate