missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl(2,4,6-trimethylbenzoyl)phosphine Oxide 98.0+%, TCI America™

Catalog No. D335825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D335825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D335825G Supplier TCI America Supplier No. D335825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75980-60-8
Molecular Formula C22H21O2P
Molecular Weight (g/mol) 348.382
MDL Number MFCD00192110
InChI Key VFHVQBAGLAREND-UHFFFAOYSA-N
PubChem CID 166480
IUPAC Name diphenylphosphoryl-(2,4,6-trimethylphenyl)methanone
SMILES CC1=CC(=C(C(=C1)C)C(=O)P(=O)(C2=CC=CC=C2)C3=CC=CC=C3)C

Specifications

Melting Point 93°C
Quantity 25 g
UN Number 3077
Formula Weight 348.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl(2,4,6-trimethylbenzoyl)phosphine Oxide
EINECSNumber (3)-4078
RTECSNumber SZ0395000
TSCA Yes