missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphényl-1-pyrénylphosphine 95,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 176°C |
| Quantity | 100 mg |
| Formula Weight | 386.43 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | Diphenyl-1-pyrenylphosphine |
Sécurité et manipulation
| TSCA | Yes |
| Recommended Storage | Réfrigérateur |