missing translation for 'onlineSavingsMsg'
Learn More

Dioctadecyl Ether 95.0+%, TCI America™

Catalog No. O001625G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O001625G 25 g
O0016250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O001625G Supplier TCI America Supplier No. O001625G

Chemical Identifiers

CAS 6297-03-6
Molecular Formula C36H74O
Molecular Weight (g/mol) 522.99
MDL Number MFCD00048498
InChI Key HBXWUCXDUUJDRB-UHFFFAOYSA-N
Synonym Distearyl Ether, Octadecyl Ether
PubChem CID 80526
IUPAC Name 1-octadecoxyoctadecane
SMILES CCCCCCCCCCCCCCCCCCOCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 63°C
Boiling Point 131°C
Quantity 25 g
Formula Weight 522.99
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dioctadecyl Ether