missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphenylphosphine, 96%

Catalog No. AC310100050
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC310100050 5 mL Glass bottle
AC310100250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC310100050 Supplier Thermo Scientific Chemicals Supplier No. 310100050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 672-66-2 | C8H11P | 138.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 672-66-2
Molecular Formula C8H11P
Molecular Weight (g/mol) 138.15
MDL Number MFCD00008509
InChI Key HASCQPSFPAKVEK-UHFFFAOYSA-N
Synonym dimethylphenylphosphine, dimethyl phenyl phosphine, dimethyl phenyl phosphane, me2pph, phenyldimethylphosphine, pme2ph, phosphine, dimethylphenyl, pubchem6420, dimethyl phenylphosphine
PubChem CID 69597
ChEBI CHEBI:30671
IUPAC Name dimethyl(phenyl)phosphane
SMILES CP(C)C1=CC=CC=C1

Specifications

Color Colorless
Density 0.9710g/mL
Boiling Point 74.0°C to 75.0°C (12.0 mmHg)
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5620 to 1.5640
Linear Formula C6H5P(CH3)2
Quantity 5 mL
Specific Gravity 0.971
Formula Weight 138.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dimethylphenylphosphine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-595-3

RUO – Research Use Only