missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylphenacylsulfonium Tetrafluoroborate 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 170°C |
| Color | White |
| Quantity | 1 g |
| UN Number | 1759 |
| Formula Weight | 268.08 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dimethylphenacylsulfonium Tetrafluoroborate |
Sécurité et manipulation
| TSCA | No |