Learn More
Dimethylglyoxime, 1% Alcoholic, for Nickel, Certified, LabChem™
$51.46 - $51.46
Chemical Identifiers
| CAS | 95-45-4 |
|---|---|
| Molecular Formula | C4H8N2O2 |
| Molecular Weight (g/mol) | 116.12 |
| MDL Number | MFCD00002117 |
| InChI Key | JGUQDUKBUKFFRO-GGWOSOGESA-N |
| Synonym | biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky |
| PubChem CID | 135399895 |
| IUPAC Name | (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine |
| SMILES | C\C(=N/O)\C(\C)=N\O |
Chemical Identifiers
| 95-45-4 | |
| 116.12 | |
| JGUQDUKBUKFFRO-GGWOSOGESA-N | |
| 135399895 | |
| C\C(=N/O)\C(\C)=N\O |
| C4H8N2O2 | |
| MFCD00002117 | |
| biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky | |
| (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine |
Specifications
| Yellow | |
| 78°C | |
| 1,89.1,4.95,4.95 | |
| CH3C(=NOH)C(=NOH)CH3 | |
| UN1170 | |
| biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky | |
| JGUQDUKBUKFFRO-GGWOSOGESA-N | |
| (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine | |
| 135399895 | |
| Certified | |
| Poly Bottle | |
| Passes Test | |
| 125 mL | |
| Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility or the unborn child (oral).
Causes damage to organs (central nervous system, optic nerve, liver, kidneys).
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/attention.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish.
Store locked up in a well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature