missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime, 1% Alcoholic, for Nickel, Certified, LabChem™

Catalog No. LC136007
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC136007 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC136007 Supplier LabChem Supplier No. LC136007

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-45-4 , 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002117
InChI Key JGUQDUKBUKFFRO-GGWOSOGESA-N
Synonym biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
PubChem CID 135399895  
IUPAC Name (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine
SMILES C\C(=N/O)\C(\C)=N\O

Specifications

Color Yellow
Boiling Point 78°C
Melting Point -115°C
CAS Max % 1, 89.1, 4.95, 4.95
Linear Formula CH3C(=NOH)C(=NOH)CH3
UN Number UN1170
Density 790kg/m3
Solubility Information Soluble in water
Formula Weight 116.12
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 790kg/m3
Suitability Passes Test
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases
Quantity 125 mL
Flash Point 13°C
Physical Form Liquid
Chemical Name or Material Dimethylglyoxime, 1% Alcoholic, for Nickel
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility or the unborn child (oral).
Causes damage to organs (central nervous system, optic nerve, liver, kidneys).
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/attention.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish.
Store locked up in a well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature