missing translation for 'onlineSavingsMsg'
Learn More

Bromure de diméthyldioctylammonium 97,0+%, TCI America™

Catalog No. D27545G
Change view
Click to view available options
Quantité:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
D27545G 5 g
D275425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27545G Supplier TCI America Supplier No. D27545G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3026-69-5
Formule moléculaire C18H40BrN
Poids moléculaire (g/mol) 350.429
Numéro MDL MFCD00082931
Clé InChI APTVNWGLSRAOFJ-UHFFFAOYSA-M
PubChem CID 76408
Nom de l’IUPAC diméthyl(dioctyl)azanium; bromure
SOURIRES CCCCCCCC[N+](C)(C)CCCCCCCC.[Br-]

Specifications

Nom ou substance chimique Dimethyldioctylammonium Bromide
Couleur Blanc-jaune
Quantité 5 g
Poids de formule 350.43
Pourcentage de pureté ≥97,0% (T)
Forme physique Poudre cristalline
EINECSNumber (9)-1971
TSCA Non