missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
$152.98 - $826.24
Specifications
| Chemical Name or Material | Dimethylamine |
|---|---|
| Color | Yellow |
| Name Note | 2M solution in THF |
| CAS | 124-40-3 , 109-99-9 |
| Molecular Formula | C2H7N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC388381000
|
Thermo Scientific Chemicals
388381000 |
100 mL |
Each for $152.98
|
|
|||||
|
AC388388000
|
Thermo Scientific Chemicals
388388000 |
800 mL |
Each for $826.24
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Dimethylamine | |
| 2M solution in THF | |
| C2H7N | |
| MFCD00008288 | |
| 0.8500g/mL | |
| dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution | |
| ROSDSFDQCJNGOL-UHFFFAOYSA-N | |
| N-methylmethanamine | |
| 674 | |
| 45.08 | |
| 0.85 | |
| Solution |
RUO – Research Use Only