accessibility menu, dialog, popup

UserName

H3CNHCH3

CAS RN 124-40-3

IUPAC Name: dimethylamine
Synonyms: dimethylamine dimethylamine (anhydrous) dimethylamine, in aqueous solution dimethylamine anhydrous hnme2 dimethylamine aqueous solution ccris 981 rcra waste number U092 dimethylamine solution dimethylamine anhydrous (dot)
Molecular Weight (g/mol): 45.09
Molecular Formula: C2H7N
InChi Key: ROSDSFDQCJNGOL-UHFFFAOYSA-N
SMILES: CNC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)