missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Yellow Indicator, Solstice

Catalog No. 60046204
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046204 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60046204 Supplier Solstice Supplier No. 33129100G

missing translation for 'validMainframeMsg'

Indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 60-11-7
Molecular Formula C14H15N3
Molecular Weight (g/mol) 225.30
MDL Number MFCD00008308
InChI Key JCYPECIVGRXBMO-UHFFFAOYSA-N
PubChem CID 6053
ChEBI CHEBI:17903
IUPAC Name N,N-dimethyl-4-phenyldiazenylaniline
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 111°C
Odor Odourless
Packaging Glass Bottle
Linear Formula C6H5N=NC6H4N(CH3)2
UN Number UN2811
Beilstein 746016
Color Yellow
pH 5 to 7 (at 10.00 g/L 20°C)
Solubility Information Insoluble in water
Quantity 100 g
Formula Weight 225.29g/mol
Grade Indicator
Chemical Name or Material Dimethyl Yellow
Show More Show Less
Hazard Category Danger
Hazard Statement P281-P301 + P310
Precautionary Statement H301-H351
DOTInformation Class 6.1 - PG 3 - RQ - Toxic solids, organic, n.o.s. HI: all
EINECSNumber 200-455-7
RTECSNumber BX7350000