missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Trisulfide 98.0+%, TCI America™

Catalog No. D34185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34185G 5 g
D341825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34185G Supplier TCI America Supplier No. D34185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3658-80-8
Molecular Formula C2H6S3
Molecular Weight (g/mol) 126.25
MDL Number MFCD00039808
InChI Key YWHLKYXPLRWGSE-UHFFFAOYSA-N
Synonym dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts
PubChem CID 19310
ChEBI CHEBI:4614
IUPAC Name (methyltrisulfanyl)methane
SMILES CSSSC

Specifications

Melting Point -68°C
Color Yellow
Boiling Point 170°C
Quantity 5 g
UN Number 1993
Formula Weight 126.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Trisulfide