missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl trans-2,3-Dibromobutenedioate 98.0+%, TCI America™

Catalog No. D40735G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40735G Supplier TCI America Supplier No. D40735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 116631-94-8
Molecular Formula C6H6Br2O4
Molecular Weight (g/mol) 301.918
InChI Key XMUOGQHVYZDUCF-ONEGZZNKSA-N
Synonym trans-2,3-Dibromobutenedioic Acid Dimethyl Ester, Dimethyl 2,3-Dibromofumarate, 2,3-Dibromofumaric Acid Dimethyl Ester
PubChem CID 11087957
IUPAC Name dimethyl (E)-2,3-dibromobut-2-enedioate
SMILES COC(=O)C(=C(C(=O)OC)Br)Br

Specifications

Melting Point 40°C
Color White-Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 301.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl trans-2,3-Dibromobutenedioate
TSCA No
Recommended Storage Refrigerator