missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide, USP, 99.9%, Spectrum™ Chemical
D1258, 67-68-5, C2H6OS
$932.40 - $932.40
Chemical Identifiers
| CAS | 67-68-5 |
|---|---|
| Molecular Formula | C2H6OS |
| Molecular Weight (g/mol) | 78.13 |
| MDL Number | MFCD00002089 |
| InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
| IUPAC Name | methanesulfinylmethane |
| SMILES | CS(C)=O |