missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, USP, 99.9%, Spectrum™ Chemical

Catalog No. 18602241
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602241 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18602241 Supplier Spectrum Chemical Mfg Cor Supplier No. D1258100ML

D1258, 67-68-5, C2H6OS

Spectrum™ Chemical Dimethyl Sulfoxide, USP is also known as DMSO. It is used topically to decrease pain and speed the healing of wounds, burns, and muscle and skeletal injuries. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Refractive Index 1.4755-1.4775
Quantity 100 mL
Specific Gravity 1.095-1.101
Percent Purity 99.9%
Grade USP