missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide, PHOTREX™ Reagent, J.T. Baker™
Description
- For Spectrophotometry
Specifications
Specifications
| Quantity | 4 L |
| Flash Point | 89.00°C |
| Packaging | Narrow Mouth PVC-coated Amber Bottle |
| Specific Gravity | 1L = 1.10kg |
| Formula Weight | 78.13 |
| Grade | PHOTREX™ Reagent |
| Chemical Name or Material | Dimethyl Sulfoxide |