missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide Molecular Biology MP Biomedicals

Catalog No. ICN19481980
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19481980 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19481980 Supplier MP Biomedicals Inc Supplier No. 0219481980

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Density 1.104 g/cm3 at 20 °C
Quantity 100 mL
Formula Weight 78.13
Grade Molecular Biology
Chemical Name or Material Dimethyl Sulfoxide