missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide 99.0+%, TCI America™

Catalog No. D5293500ML
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5293500ML 500 mL
D5293100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5293500ML Supplier TCI America Supplier No. D5293500ML
Only null left

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Color Colorless
Quantity 500 mL
Physical Form Clear Liquid at 20°C
Boiling Point 189°C
Formula Weight 78.13
Percent Purity ≥99.0% (GC)
Chemical Name or Material Dimethyl Sulfoxide [for Spectrophotometry]
EINECSNumber (2)-1553
RTECSNumber PV6210000
TSCA Yes