missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide 99.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 100 mL |
| Physical Form | Clear Liquid at 20°C |
| Boiling Point | 189°C |
| Formula Weight | 78.13 |
| Percent Purity | ≥99.0% (GC) |
| Chemical Name or Material | Dimethyl Sulfoxide [for Spectrophotometry] |
Safety and Handling
| EINECSNumber | (2)-1553 |
| RTECSNumber | PV6210000 |
| TSCA | Yes |