missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, CHROMASOLV™ Plus, for HPLC, ≥99.7%, Solstice

Catalog No. 6002583
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002583 6 x 1 L
6002578 1 L
6002579 2.5 L
60046492 4 x 2.5 L
6002581 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.
Catalog No. 6002583 Supplier Solstice Supplier No. 348696X1L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

CHROMASOLV™ Plus, for HPLC,≥99.7%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Melting Point 16°C to 19°C
Density 1.10g/mL
Quantity 6 x 1 L
Refractive Index n20/D 1.479
Linear Formula (CH3)2SO
Vapor Pressure 0.42mmHg (20°C)
Beilstein 506008
Boiling Point 189°C
IUPAC Name methylsulfinylmethane
Formula Weight 78.13g/mol
Percent Purity ≥99.7%
Grade CHROMASOLV™ Plus
Chemical Name or Material Dimethyl sulfoxide, for HPLC
Show More Show Less
EINECSNumber 200-664-3
RTECSNumber PV6210000