missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023061
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023061 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023061 Supplier Merck Emd Millipore Supplier No. RTC00010320ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

Melting Point 16°C to 19°C (literature)
CAS 67-68-5
Density 1.10 g/mL (literature)
Quantity 20 mL
Flash Point 87°C (Not Applicable)
Refractive Index n20/D 1.479 (literature)
Molecular Formula C2H6OS
Vapor Pressure 0.42 mmHg (20°C)
Linear Formula (CH3)2 SO
MDL Number MFCD00002089
Synonym Dimethyl sulfoxide; DMSO
Boiling Point 189°C (literature)
Formula Weight 78.13
Grade Reference Material
Show More Show Less