missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02002735
Change view
Click to view available options
Quantity:
500 mL
19 L
Packaging:
HDPE Pressure Bottom Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002735 500 mL HDPE Pressure Bottom Poly Bottle
02002728 19 L Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002735 Supplier Avantor Supplier No. 922406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Quantity 500 mL
Flash Point 89.00°C
Packaging HDPE Pressure Bottom Poly Bottle
Specific Gravity 1L = 1.10kg
IUPAC Name methanesulfinylmethane
Formula Weight 78.13
Grade Meets ACS reagent chemical requirements
Chemical Name or Material Dimethyl Sulfoxide