missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, ACS Reagent (DMSO), Reagents

Catalog No. 01338004
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01338004 4 L Poly Bottle
01338003 4 L Glass Bottle
01338002 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01338004 Supplier Labchem Inc Supplier No. C11966504A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dimethyl Sulfoxide, ACS Reagent (DMSO), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym DMSO, Methylsulfinylmethane
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.5°C
Color Colorless
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.101
Boiling Point 189°C
Grade ACS
Chemical Name or Material Dimethyl Sulfoxide, ACS Reagent (DMSO)