missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, ≥99.5%, Reagent Grade, Solstice

Catalog No. 60047971
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047971 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60047971 Supplier Solstice Supplier No. D5879500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent Grade,≥99.5%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.5°C
Color Colorless
Density 1.10g/mL
Quantity 500 mL
Explosion Limit 42% (63°F)
Flash Point 87°C (188.6°F)
Ignition Point 573°F
Odor Characteristic
Packaging Poly Bottle
Physical Form Liquid
Vapor Pressure 0.42mmHg (20°C)
Linear Formula (CH3)2SO
Beilstein 506008
Boiling Point 189°C
Formula Weight 78.13g/mol
Percent Purity ≥99.5%
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
RTECSNumber 200-664-3