missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl succinate, 98%

Catalog No. AAA1256530
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1256530 250 g
AAA1256522 100 g
AAA125650B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1256530 Supplier Thermo Scientific Chemicals Supplier No. A1256530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-65-0 | C6H10O4 | 146.142 g/mol

Dimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.

Solubility
Soluble in water. (122,9 g/L at 20°C). soluble in acetone, ethanol, very soluble in ethyl ether.

Notes
Store at room temperature. Ensure good ventilation.

Chemical Identifiers

CAS 106-65-0
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00008466
InChI Key MUXOBHXGJLMRAB-UHFFFAOYSA-N
Synonym dimethyl succinate, methyl succinate, dimethylsuccinate, butanedioic acid, dimethyl ester, succinic acid, dimethyl ester, dbe-4 dibasic ester, methyl butanedioate, succinic acid dimethyl ester, butanedioic acid, 1,4-dimethyl ester, fema no. 2396
PubChem CID 7820
IUPAC Name dimethyl butanedioate
SMILES COC(=O)CCC(=O)OC

Specifications

Melting Point 18°C to 19°C
Density 1.119
Boiling Point 196°C
Flash Point 85°C (185°F)
Refractive Index 1.419
Quantity 250 g
Beilstein 956776
Merck Index 14,8869
Solubility Information Soluble in water. (122,9g/L at 20°C). soluble in acetone,ethanol,very soluble in ethyl ether.
Formula Weight 146.14
Percent Purity ≥98%
Chemical Name or Material Dimethyl succinate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 203-419-9
RTECSNumber WM7675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only