missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl suberate, 99%
CAS: 1732-09-8 | C10H18O4 | 202.25 g/mol
$89.14 - $252.44
Chemical Identifiers
| CAS | 1732-09-8 |
|---|---|
| Molecular Formula | C10H18O4 |
| Molecular Weight (g/mol) | 202.25 |
| MDL Number | MFCD00008471 |
| InChI Key | LNLCRJXCNQABMV-UHFFFAOYSA-N |
| Synonym | dimethyl suberate, octanedioic acid, dimethyl ester, suberic acid, dimethyl ester, octanedioic acid dimethyl ester, suberic acid dimethyl ester, octanedioic acid dimethyl, 1,8-dimethyl octanedioate, octanedioic acid, 1,8-dimethyl ester, dimethyl octane-1,8-dioate |
| PubChem CID | 15611 |
| ChEBI | CHEBI:81345 |
| IUPAC Name | dimethyl octanedioate |
| SMILES | COC(=O)CCCCCCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAB2268114
|
N/A | 25 g |
Each for $89.14
|
|
|||||
|
AAB2268122
|
N/A | 100 g |
Each for $252.44
|
|
|||||
Description
Dimethyl suberate was used in the preparation of [8-2H2] and [7-2H2]oleates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethyl suberate was used in the preparation of [8-2H2] and [7-2H2]oleates.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| C10H18O4 | |
| MFCD00008471 | |
| dimethyl suberate, octanedioic acid, dimethyl ester, suberic acid, dimethyl ester, octanedioic acid dimethyl ester, suberic acid dimethyl ester, octanedioic acid dimethyl, 1,8-dimethyl octanedioate, octanedioic acid, 1,8-dimethyl ester, dimethyl octane-1,8-dioate | |
| CHEBI:81345 | |
| COC(=O)CCCCCCC(=O)OC |
Specifications
| 1732-09-8 | |
| 266°C to 268°C | |
| C10H18O4 | |
| MFCD00008471 | |
| dimethyl suberate, octanedioic acid, dimethyl ester, suberic acid, dimethyl ester, octanedioic acid dimethyl ester, suberic acid dimethyl ester, octanedioic acid dimethyl, 1,8-dimethyl octanedioate, octanedioic acid, 1,8-dimethyl ester, dimethyl octane-1,8-dioate | |
| COC(=O)CCCCCCC(=O)OC | |
| 202.25 | |
| CHEBI:81345 | |
| 99% |
| 1.016 | |
| >110°C (230°F) | |
| 1.432 | |
| 1780054 | |
| LNLCRJXCNQABMV-UHFFFAOYSA-N | |
| dimethyl octanedioate | |
| 15611 | |
| 202.25 | |
| Dimethyl suberate |
Safety and Handling
EINECSNumber : 257-059-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only