accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 1732-09-8

IUPAC Name: 1,8-dimethyl octanedioate
Synonyms: octanedioic acid, dimethyl ester
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: LNLCRJXCNQABMV-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCC(=O)OC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)