missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl suberate, 99%

Catalog No. AAB2268114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2268114 25 g
AAB2268122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2268114 Supplier Thermo Scientific Chemicals Supplier No. B2268114

CAS: 1732-09-8 | C10H18O4 | 202.25 g/mol

Dimethyl suberate was used in the preparation of [8-2H2] and [7-2H2]oleates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl suberate was used in the preparation of [8-2H2] and [7-2H2]oleates.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1732-09-8
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00008471
InChI Key LNLCRJXCNQABMV-UHFFFAOYSA-N
Synonym dimethyl suberate, octanedioic acid, dimethyl ester, suberic acid, dimethyl ester, octanedioic acid dimethyl ester, suberic acid dimethyl ester, octanedioic acid dimethyl, 1,8-dimethyl octanedioate, octanedioic acid, 1,8-dimethyl ester, dimethyl octane-1,8-dioate
PubChem CID 15611
ChEBI CHEBI:81345
IUPAC Name dimethyl octanedioate
SMILES COC(=O)CCCCCCC(=O)OC

Specifications

Density 1.016
Boiling Point 266°C to 268°C
Flash Point >110°C (230°F)
Refractive Index 1.432
Quantity 25 g
Beilstein 1780054
Formula Weight 202.25
Percent Purity 99%
Chemical Name or Material Dimethyl suberate
EINECSNumber 257-059-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only