missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Octafluoroadipate 97.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Boiling Point | 123°C |
| Quantity | 1 g |
| Formula Weight | 318.12 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dimethyl Octafluoroadipate |
Sécurité et manipulation
| TSCA | No |