missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl maleate, 96%, Thermo Scientific™

Catalog No. AC218460051
Change view
Click to view available options
5l, Plastic drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC218460051 5l, Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC218460051 Supplier Thermo Scientific Chemicals Supplier No. 218460051

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 624-48-6
Density 1.1500g/mL
Boiling Point 204.0°C to 205.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Plastic drum
Molecular Formula C6H8O4
Linear Formula CH3O2CCH=CHCO2CH3
Refractive Index 1.4410 to 1.4430
Quantity 5L
MDL Number MFCD00008459
Beilstein 02,751
Specific Gravity 1.15
Synonym dimethyl maleate, maleic acid, dimethyl ester, methyl maleate, sipomer dmm, maleic acid dimethyl ester, dimethyl cis-butenedioate, 2-butenedioic acid z-, dimethyl ester, dimethyl cis-ethylenedicarboxylate, 2-butenedioic acid 2z-, dimethyl ester, 2-butenedioic acid, dimethyl ester, z
Solubility Information Solubility in water: 80g/L (25°C)
InChI Key LDCRTTXIJACKKU-ARJAWSKDSA-N
SMILES COC(=O)C=CC(=O)OC
IUPAC Name dimethyl (Z)-but-2-enedioate
Molecular Weight (g/mol) 144.13
PubChem CID 5271565
Viscosity 4 mPa.s (20°C)
ChEBI CHEBI:35460
Formula Weight 144.13
Percent Purity 96%
Chemical Name or Material Dimethyl maleate, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
EINECSNumber 210-848-5
RTECSNumber EM6300000
TSCA TSCA

RUO – Research Use Only