missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl L-Aspartate Hydrochloride 98.0+%, TCI America™

Catalog No. A15065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15065G 5 g
A150625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15065G Supplier TCI America Supplier No. A15065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity laboratory reagent with ≥98.0% purity for use as an amino acid derivative in peptide chemistry.

  • Chemical Name: Dimethyl L-Aspartate Hydrochloride
  • Molecular Weight: 197.62 g/mol
  • Appearance: White to almost white powder to crystal
  • Melting Point: 113.0°C to 118.0°C
  • Purity (qNMR): ≥98.0%
  • Specific Rotation: +11.0° to +13.0° (C=1.4, H2O)
  • Storage: Room temperature, store under inert gas, protect from moisture

Chemical Identifiers

CAS 32213-95-9
Molecular Formula C6H12ClNO4
Molecular Weight (g/mol) 197.615
MDL Number MFCD00038878
InChI Key PNLXWGDXZOYUKB-WCCKRBBISA-N
Synonym h-asp ome-ome hcl, l-aspartic acid dimethyl ester hydrochloride, dimethyl l-aspartate hydrochloride, h-asp ome-ome.hcl, methyl aspartic acid hydrochloride, s-dimethyl 2-aminosuccinate hydrochloride, aspartic acid dimethyl ester hydrochloride, l-aspartic acid dimethyl ester hydro-chloride, s-aminosuccinic acid dimethyl ester hydrochloride, dimethyl aspartic acid hydrochloride
PubChem CID 2734892
IUPAC Name dimethyl (2S)-2-aminobutanedioate;hydrochloride
SMILES COC(=O)CC(C(=O)OC)N.Cl

Specifications

Melting Point 116°C
Color White
Quantity 5 g
Formula Weight 197.62
Percent Purity ≥98.0% (N,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl L-Aspartate Hydrochloride