missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Diselenide 98.0+%, TCI America™

Catalog No. D32901G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D32901G Supplier TCI America Supplier No. D32901G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7101-31-7
Molecular Formula C2H6Se2
Molecular Weight (g/mol) 188.012
InChI Key VLXBWPOEOIIREY-UHFFFAOYSA-N
Synonym dimethyl diselenide, dimethyldiselenide, methyl diselenide, diselenide, dimethyl, ch3se 2, methyldiselanyl methane, diselenobismethane, 1,2-dimethyldiselane, acmc-209ogy, methyl diselenide, 8ci
PubChem CID 23496
IUPAC Name (methyldiselanyl)methane
SMILES C[Se][Se]C

Specifications

Melting Point -82°C
Boiling Point 155°C
Quantity 1 g
UN Number 3440
Formula Weight 187.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Diselenide