missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl cis-stilbene-4,4'-dicarboxylate
CAS: 143130-82-9 | C18H16O4 | 296.322 g/mol
Supplier: Thermo Scientific Chemicals L1114203
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Dimethyl cis-stilbene-4,4'-dicarboxylate | |
| 112°C to 114°C | |
| MFCD00082669 | |
| 2386934 | |
| JOODVYOWCWQPMV-ARJAWSKDSA-N | |
| methyl 4-[(Z)-2-(4-methoxycarbonylphenyl)ethenyl]benzoate | |
| 7556518 |
| 143130-82-9 | |
| C18H16O4 | |
| 1 g | |
| dimethyl cis-stilbene-4,4'-dicarboxylate, dimethyl cis-stilbene-4,4/'-dicarboxylate, z-dimethyl 4,4'-ethene-1,2-diyl dibenzoate, z-4,4'-stilbenedicarboxylic acid dimethyl ester, methyl 4-z-2-4-methoxycarbonylphenyl ethenyl benzoate, methyl 4-z-2-4-methoxycarbonyl phenyl ethenyl benzoate | |
| COC(=O)C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C(=O)OC | |
| 296.322 | |
| 296.33 |
Chemical Identifiers
| 143130-82-9 | |
| 296.322 | |
| JOODVYOWCWQPMV-ARJAWSKDSA-N | |
| 7556518 | |
| COC(=O)C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C(=O)OC |
| C18H16O4 | |
| MFCD00082669 | |
| dimethyl cis-stilbene-4,4'-dicarboxylate, dimethyl cis-stilbene-4,4/'-dicarboxylate, z-dimethyl 4,4'-ethene-1,2-diyl dibenzoate, z-4,4'-stilbenedicarboxylic acid dimethyl ester, methyl 4-z-2-4-methoxycarbonylphenyl ethenyl benzoate, methyl 4-z-2-4-methoxycarbonyl phenyl ethenyl benzoate | |
| methyl 4-[(Z)-2-(4-methoxycarbonylphenyl)ethenyl]benzoate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only