accessibility menu, dialog, popup

UserName

CAS RN 143130-82-9

IUPAC Name: methyl 4-{2-[4-(methoxycarbonyl)phenyl]ethenyl}benzoate
Synonyms: methyl 4-{2-[4-(methoxycarbonyl)phenyl]ethenyl}benzoate
Molecular Weight (g/mol): 296.32
Molecular Formula: C18H16O4
InChi Key: JOODVYOWCWQPMV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=CC2=CC=C(C=C2)C(=O)OC)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results