missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl cis-stilbene-4,4'-dicarboxylate

Catalog No. p-7045558
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL11142MD 250 mg
AAL1114203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL11142MD Supplier Thermo Scientific Chemicals Supplier No. L11142MD
Only null left

CAS: 143130-82-9 | C18H16O4 | 296.322 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 143130-82-9
Molecular Formula C18H16O4
Molecular Weight (g/mol) 296.322
MDL Number MFCD00082669
InChI Key JOODVYOWCWQPMV-ARJAWSKDSA-N
Synonym dimethyl cis-stilbene-4,4'-dicarboxylate, dimethyl cis-stilbene-4,4/'-dicarboxylate, z-dimethyl 4,4'-ethene-1,2-diyl dibenzoate, z-4,4'-stilbenedicarboxylic acid dimethyl ester, methyl 4-z-2-4-methoxycarbonylphenyl ethenyl benzoate, methyl 4-z-2-4-methoxycarbonyl phenyl ethenyl benzoate
PubChem CID 7556518
IUPAC Name methyl 4-[(Z)-2-(4-methoxycarbonylphenyl)ethenyl]benzoate
SMILES COC(=O)C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C(=O)OC

Specifications

Melting Point 112°C to 114°C
Quantity 250 mg
Beilstein 2386934
Formula Weight 296.33
Chemical Name or Material Dimethyl cis-stilbene-4,4'-dicarboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only