missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl cis-4-Cyclohexene-1,2-dicarboxylate 97.0+%, TCI America™

Catalog No. D37035G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D37035G Supplier TCI America Supplier No. D37035G
Only null left

Chemical Identifiers

CAS 4841-84-3
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
MDL Number MFCD00209581
InChI Key DVVAGRMJGUQHLI-OCAPTIKFSA-N
Synonym dimethyl cis-4-cyclohexene-1,2-dicarboxylate, dimethyl cis-1,2,3,6-tetrahydrophthalate, cis-dimethyl cyclohex-4-ene-1,2-dicarboxylate, dimethylcis-4-cyclohexene-1,2-dicarboxylate, 1r, 6s-methyl 6-methoxycarbonyl-3-cyclohexenecarboxylate, 4-cyclohexene-1alpha,2alpha-dicarboxylic acid dimethyl ester, 1,2-dimethyl 1r,2s-cyclohex-4-ene-1,2-dicarboxylate, 4-cyclohexene-1,2-dicarboxylicacid, dimethyl ester, 1r,2s-rel
PubChem CID 10867283
IUPAC Name dimethyl (1R,2S)-cyclohex-4-ene-1,2-dicarboxylate
SMILES COC(=O)C1CC=CCC1C(=O)OC

Specifications

Color Yellow
Boiling Point 142°C
Quantity 5 g
Formula Weight 198.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl cis-4-Cyclohexene-1,2-dicarboxylate
EINECSNumber (3)-2437
TSCA No