missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl cis-1,2-cyclopropanedicarboxylate, 97+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 255340250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC255340250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

826-34-6
158.16
JBVOSZYUSFDYIN-SYDPRGILSA-N
642979
COC(=O)C1CC1C(=O)OC
C7H10O4
MFCD00062805
dimethyl cis-1,2-cyclopropanedicarboxylate, dimethyl 1r,2s-cyclopropane-1,2-dicarboxylate, cis-dimethyl cyclopropane-1,2-dicarboxylate, dimethylcis-1,2-cyclopropanedicarboxylate, cyclopropane-1,2-dicarboxylic acid dimethyl ester, z, 1,2-cyclopropanedicarboxylic acid, dimethyl ester, 1r,2s-rel, 1,2-dimethyl 1r,2s-cyclopropane-1,2-dicarboxylate, 1r,2s-dimethyl cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, dimethyl ester, 1r,2s, 1,2-cyclopropanedicarboxylicacid, 1,2-dimethyl ester, 1r,2s-rel
dimethyl (1S,2R)-cyclopropane-1,2-dicarboxylate

Specifications

Dimethyl cis-1,2-cyclopropanedicarboxylate
1.1500g/mL
73°C
97% min. (GC)
C7H10O4
25g
1.15
Solubility in water: insoluble. Other solubilities: soluble in all organic solvents
COC(=O)C1CC1C(=O)OC
158.16
158.16
826-34-6
112.0°C (18.0 mmHg)
Authentic
Glass bottle
1.4445 to 1.4465
MFCD00062805
dimethyl cis-1,2-cyclopropanedicarboxylate, dimethyl 1r,2s-cyclopropane-1,2-dicarboxylate, cis-dimethyl cyclopropane-1,2-dicarboxylate, dimethylcis-1,2-cyclopropanedicarboxylate, cyclopropane-1,2-dicarboxylic acid dimethyl ester, z, 1,2-cyclopropanedicarboxylic acid, dimethyl ester, 1r,2s-rel, 1,2-dimethyl 1r,2s-cyclopropane-1,2-dicarboxylate, 1r,2s-dimethyl cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, dimethyl ester, 1r,2s, 1,2-cyclopropanedicarboxylicacid, 1,2-dimethyl ester, 1r,2s-rel
JBVOSZYUSFDYIN-SYDPRGILSA-N
dimethyl (1S,2R)-cyclopropane-1,2-dicarboxylate
642979
97+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...