missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl cis-1,2-Cyclohexanedicarboxylate 96.0+%, TCI America™

Catalog No. D37025G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37025G 5 g
D37021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37025G Supplier TCI America Supplier No. D37025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1687-29-2
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
MDL Number MFCD00043768,MFCD21602391
InChI Key AIACXWOETVLBIA-UHFFFAOYNA-N
Synonym cis-1,2-Cyclohexanedicarboxylic Acid Dimethyl Ester
PubChem CID 642993
IUPAC Name 1,2-dimethyl cyclohexane-1,2-dicarboxylate
SMILES COC(=O)C1CCCCC1C(=O)OC

Specifications

Color Yellow
Boiling Point 111°C
Quantity 5 g
Formula Weight 200.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl cis-1,2-Cyclohexanedicarboxylate