missing translation for 'onlineSavingsMsg'
Learn More

Diméthyl chlorophosphate, 96%

Numéro de catalogue. p-3770863
Modifier l'affichage
Click to view available options
Quantité:
5 g
Emballage:
Bouteille en verre
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC426140050 5 g Bouteille en verre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC426140050 Fournisseur Thermo Scientific Chemicals Code fournisseur 426140050

CAS : 813-77-4 | C2H6ClO3P | 144,49 g/mol

Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.

Identifiants chimiques

CAS 813-77-4
Formule moléculaire C2H6ClO3P
Poids moléculaire (g/mol) 144.49
Numéro MDL MFCD00117904
Clé InChI NGFFLHMFSINFGB-UHFFFAOYSA-N
Synonyme dimethyl phosphorochloridate, dimethyl chlorophosphate, phosphorochloridic acid, dimethyl ester, methyl phosphorochloridate, dimethylchlorophosphate, dimethyl chlorophosphonate, chlorodimethoxyphosphine oxide, chlorodimethylphosphate, dimethyl_chlorophosphate, pubchem19321
PubChem CID 101863
SOURIRES COP(Cl)(=O)OC

Spécifications

Nom ou substance chimique Dimethyl chlorophosphate
Densité 1.3400g/mL
Point d’ébullition 80 °C (25,0 mmHg)
Point d’éclair >110 °C
Fourchette de pourcentages d’essai 95% min. (Argentométrie)
Emballage Bouteille en verre
Formule linéaire (CH3O)2POCl
Quantité 5 g
Gravité spécifique 1.34
Nom de l’IUPAC Diméthylphosphorochloridate
Poids de formule 144.49
Pourcentage de pureté 96%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if swallowed.
Causes severe skin burns and eye damage.
Fatal in contact with skin.
Fatal if inhaled.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Usage réservé à la recherche