missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl azelate, 97%
CAS: 1732-10-1 | C11H20O4 | 216.28 g/mol
$106.19 - $1,049.79
Chemical Identifiers
| CAS | 1732-10-1 |
|---|---|
| Molecular Formula | C11H20O4 |
| Molecular Weight (g/mol) | 216.28 |
| MDL Number | MFCD00025898 |
| InChI Key | DRUKNYVQGHETPO-UHFFFAOYSA-N |
| Synonym | dimethyl azelate, nonanedioic acid, dimethyl ester, methyl azelate, azelaic acid dimethyl ester, azelaic acid, dimethyl ester, 1,9-dimethyl nonanedioate, unii-29185k7oei, nonanedioic acid, 1,9-dimethyl ester, nonanedioic acid dimethyl ester, dimethyl nonane-1,9-dioate |
| PubChem CID | 15612 |
| SMILES | COC(=O)CCCCCCCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAL1745806
|
N/A | 5 g |
Each for $106.19
|
|
|||||
|
AAL1745814
|
N/A | 25 g |
Each for $377.78
|
|
|||||
|
AAL1745822
|
N/A | 100 g |
Each for $1,049.79
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C11H20O4 | |
| MFCD00025898 | |
| dimethyl azelate, nonanedioic acid, dimethyl ester, methyl azelate, azelaic acid dimethyl ester, azelaic acid, dimethyl ester, 1,9-dimethyl nonanedioate, unii-29185k7oei, nonanedioic acid, 1,9-dimethyl ester, nonanedioic acid dimethyl ester, dimethyl nonane-1,9-dioate | |
| COC(=O)CCCCCCCC(=O)OC |
Specifications
| 1732-10-1 | |
| 1.007 | |
| >110°C (230°F) | |
| 1.437 | |
| MFCD00025898 | |
| 14,905 | |
| DRUKNYVQGHETPO-UHFFFAOYSA-N | |
| 1,9-dimethyl nonanedioate | |
| 15612 | |
| 97% |
| 18°C | |
| 156°C (20 mmHg) | |
| C11H20O4 | |
| CH3OOC(CH2)7COOCH3 | |
| 1710125 | |
| dimethyl azelate, nonanedioic acid, dimethyl ester, methyl azelate, azelaic acid dimethyl ester, azelaic acid, dimethyl ester, 1,9-dimethyl nonanedioate, unii-29185k7oei, nonanedioic acid, 1,9-dimethyl ester, nonanedioic acid dimethyl ester, dimethyl nonane-1,9-dioate | |
| COC(=O)CCCCCCCC(=O)OC | |
| 216.28 | |
| 216.28 | |
| Dimethyl azelate |
Safety and Handling
EINECSNumber : 217-060-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only