missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl azelate, 97%

CAS: 1732-10-1 | C11H20O4 | 216.28 g/mol

Supplier:  Thermo Scientific Chemicals L1745814

Quantity 25 g

 View more versions of this product

Catalog No. AAL1745814


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1732-10-1
216.28
DRUKNYVQGHETPO-UHFFFAOYSA-N
15612
C11H20O4
MFCD00025898
dimethyl azelate, nonanedioic acid, dimethyl ester, methyl azelate, azelaic acid dimethyl ester, azelaic acid, dimethyl ester, 1,9-dimethyl nonanedioate, unii-29185k7oei, nonanedioic acid, 1,9-dimethyl ester, nonanedioic acid dimethyl ester, dimethyl nonane-1,9-dioate
COC(=O)CCCCCCCC(=O)OC

Specifications

Dimethyl azelate
18°C
156°C (20 mmHg)
C11H20O4
CH3OOC(CH2)7COOCH3
MFCD00025898
14,905
DRUKNYVQGHETPO-UHFFFAOYSA-N
dimethyl nonanedioate
15612
97%
1732-10-1
1.007
>110°C (230°F)
1.437
25 g
1710125
dimethyl azelate, nonanedioic acid, dimethyl ester, methyl azelate, azelaic acid dimethyl ester, azelaic acid, dimethyl ester, 1,9-dimethyl nonanedioate, unii-29185k7oei, nonanedioic acid, 1,9-dimethyl ester, nonanedioic acid dimethyl ester, dimethyl nonane-1,9-dioate
COC(=O)CCCCCCCC(=O)OC
216.28
216.28
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 217-060-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions