missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl azelate, 97%

Catalog No. AAL1745806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1745806 5 g
AAL1745814 25 g
AAL1745822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1745806 Supplier Thermo Scientific Chemicals Supplier No. L1745806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1732-10-1 | C11H20O4 | 216.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1732-10-1
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.28
MDL Number MFCD00025898
InChI Key DRUKNYVQGHETPO-UHFFFAOYSA-N
Synonym dimethyl azelate, nonanedioic acid, dimethyl ester, methyl azelate, azelaic acid dimethyl ester, azelaic acid, dimethyl ester, 1,9-dimethyl nonanedioate, unii-29185k7oei, nonanedioic acid, 1,9-dimethyl ester, nonanedioic acid dimethyl ester, dimethyl nonane-1,9-dioate
PubChem CID 15612
SMILES COC(=O)CCCCCCCC(=O)OC

Specifications

Melting Point 18°C
Density 1.007
Boiling Point 156°C (20 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.437
Linear Formula CH3OOC(CH2)7COOCH3
Quantity 5 g
Beilstein 1710125
Merck Index 14,905
IUPAC Name 1,9-dimethyl nonanedioate
Formula Weight 216.28
Percent Purity 97%
Chemical Name or Material Dimethyl azelate
Show More Show Less
EINECSNumber 217-060-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only