missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Adipate 99.0+%, TCI America™

Catalog No. A016625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A016625ML 25 mL
A0166500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A016625ML Supplier TCI America Supplier No. A016625ML

Chemical Identifiers

CAS 627-93-0
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00008469
InChI Key UDSFAEKRVUSQDD-UHFFFAOYSA-N
Synonym dimethyl adipate, dimethyladipate, adipic acid dimethyl ester, hexanedioic acid, dimethyl ester, adipic acid, dimethyl ester, 1,6-dimethylhexanedioate, methyl adipate van, 1,6-dimethyl hexanedioate, dbe 6, unii-by71rx0r62
PubChem CID 12329
ChEBI CHEBI:34715
IUPAC Name dimethyl hexanedioate
SMILES COC(=O)CCCCC(=O)OC

Specifications

Color Colorless
Boiling Point 110°C
Quantity 25 mL
Formula Weight 174.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Adipate
EINECSNumber (2)-0861&(2)-0879
RTECSNumber AV1645000
TSCA Yes